Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281971.1 | NP_001268900.1 | 4 | 594 | ||
NM_001281972.1 | NP_001268901.1 | 4 | 594 | |||
NM_012314.5 | NP_036446.3 | 5 | 616 | |||
GenBank mRNA | AF002255.1 | - | 5 | 573 | ||
AF135564.1 | - | 4 | 368 | |||
AJ417554.1 | - | 3 | 485 | |||
AJ417555.1 | - | 3 | 507 | |||
AJ620616.2 | - | 3 | 478 | |||
AY102623.1 | - | 4 | 505 | |||
AY102624.1 | - | 2 | 217 | |||
AY366245.1 | - | 4 | 561 | |||
AY388472.1 | - | 4 | 565 | |||
AY789064.2 | - | 4 | 489 | |||
BC096694.1 | - | 4 | 538 | 40026979 | ||
BC101977.1 | - | 4 | 538 | 40004312 | ||
BC103693.1 | - | 4 | 538 | 40004314 | ||
BC108918.1 | - | 5 | 529 | 40017300 | ||
BC125147.1 | - | 3 | 511 | |||
BC127084.1 | - | 4 | 544 | 40120468 | ||
BC144566.1 | - | 4 | 616 | 9053094 | ||
BC171788.1 | - | 4 | 616 | 9054442 | ||
L76671.1 | - | 4 | 538 | |||
U24077.1 | - | 4 | 518 | |||
U96188.1 | - | 3 | 464 | |||
X94609.1 | - | 4 | 582 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF135560.1 | - | 2 | 464 |
Gene Symbol: KIR2DL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF135556.1 | - | 3 | 464 |