Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CITED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144885.1 | NP_001138357.1 | 4 | 899 | ||
NM_001144886.1 | NP_001138358.1 | 3 | 851 | |||
NM_001144887.1 | NP_001138359.1 | 3 | 778 | |||
NM_004143.3 | NP_004134.2 | 3 | 1199 | |||
XM_011530958.1 | XP_011529260.1 | 4 | 890 | |||
GenBank mRNA | BC004240.1 | - | 3 | 1152 | 3627230 | |
U65092.1 | - | 3 | 818 |