Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002507.3 | NP_002498.1 | 6 | 2024 | ||
GenBank mRNA | BC050309.1 | - | 6 | 2024 | 5263715 | |
M14764.1 | - | 6 | 2012 |