Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000915.3 | NP_000906.1 | 3 | 421 | ||
XM_011529238.1 | XP_011527540.1 | 4 | 558 | |||
GenBank mRNA | BC069144.1 | - | 3 | 421 | 7217004 | |
BC101841.1 | - | 3 | 419 | 8069347 | ||
BC101843.1 | - | 3 | 419 | 8069349 | ||
M25650.1 | - | 3 | 402 | |||
X03173.1 | - | 2 | 149 |