Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POMC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000939.3 | NP_000930.1 | 3 | 849 | ||
NM_001035256.2 | NP_001030333.1 | 4 | 899 | |||
NM_001319204.1 | NP_001306133.1 | 4 | 929 | |||
NM_001319205.1 | NP_001306134.1 | 3 | 879 | |||
GenBank mRNA | BC065832.1 | - | 3 | 561 | 30392189 | |
BT019918.1 | - | 2 | 475 | |||
CD106051.1 | - | 4 | 666 | |||
CD108235.1 | - | 3 | 572 | |||
CR541826.1 | - | 2 | 475 | |||
DQ884404.1 | - | 2 | 484 | |||
M38297.1 | - | 2 | 460 |