Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003083.3 | NP_003074.1 | 3 | 392 | ||
NR_030717.1 | 3 | 289 | ||||
GenBank mRNA | AK314878.1 | - | 3 | 377 | ||
BC011868.2 | - | 3 | 347 | 4545178 | ||
BP234322.1 | - | 3 | 288 | |||
BX647416.1 | - | 3 | 256 | |||
CF454397.1 | - | 3 | 392 | |||
U44755.1 | - | 3 | 376 | |||
U44898.1 | - | 3 | 358 |