Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 5 | 622 | ||
NM_001278287.1 | NP_001265216.1 | 5 | 538 | |||
NM_001278290.1 | NP_001265219.1 | 4 | 383 | |||
NM_001278291.1 | NP_001265220.1 | 3 | 252 | |||
NM_001278292.1 | NP_001265221.1 | 4 | 373 | |||
NM_001318135.1 | NP_001305064.1 | 5 | 500 | |||
NM_001318136.1 | NP_001305065.1 | 2 | 532 | |||
NM_001318137.1 | NP_001305066.1 | 3 | 252 | |||
NM_017715.3 | NP_060185.2 | 5 | 504 | |||
NM_032924.4 | NP_116313.3 | 5 | 504 | |||
XM_006716111.1 | XP_006716174.1 | 5 | 558 | |||
XM_017012582.1 | XP_016868071.1 | 5 | 572 | |||
XM_017012583.1 | XP_016868072.1 | 3 | 721 | |||
GenBank mRNA | AF193057.1 | - | 5 | 467 | ||
AF217988.1 | - | 5 | 468 | |||
AK000223.1 | - | 3 | 239 | |||
AK091984.1 | - | 2 | 504 | |||
AK297625.1 | - | 5 | 466 | |||
AK309719.1 | - | 5 | 810 | |||
AK315479.1 | - | 3 | 218 | |||
BC011887.2 | - | 5 | 428 | 3831842 | ||
BC013603.1 | - | 5 | 447 | 3865426 | ||
BC025265.1 | - | 5 | 437 | |||
BQ071498.1 | - | 5 | 334 | |||
BT006918.1 | - | 2 | 69 | |||
BX647182.1 | - | 4 | 1145 | |||
CB129643.1 | - | 3 | 208 | |||
CB988353.1 | - | 2 | 532 | |||
DA443888.1 | - | 3 | 238 | |||
DA623743.1 | - | 4 | 347 | |||
DC347416.1 | - | 4, 5 | 449 | |||
DC384519.1 | - | 5 | 504 | |||
X60153.1 | - | 3 | 177 |