Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005675.4 | NP_005666.2 | 2 | 278 | ||
XM_011530394.1 | XP_011528696.1 | 2 | 146 | |||
GenBank mRNA | AF228707.1 | - | 2 | 222 | ||
AK098780.1 | - | 2 | 146 | |||
AK315115.1 | - | 2 | 183 | |||
BC047039.1 | - | 2 | 171 | 5229172 | ||
BQ639188.1 | - | 2 | 278 | |||
CR456434.1 | - | 2 | 148 | |||
KJ534820.1 | - | 2 | 126 | |||
KJ534971.1 | - | 2 | 126 | |||
KJ535003.1 | - | 2 | 126 | |||
KJ535013.1 | - | 2 | 126 | |||
KJ535047.1 | - | 1 | 205 | |||
X96484.1 | - | 2 | 139 |
Gene Symbol: LOC102724770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724934.1 | XP_006724997.1 | 2 | 220 |