Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2AI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003509.2 | NP_003500.1 | 1 | 409 | ||
GenBank mRNA | BC112254.1 | - | 1 | 487 | 8327722 | |
BC112256.1 | - | 1 | 487 | 8327724 |