Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278720.1 | NP_001265649.1 | 6, 7 | 906 | ||
NM_001278721.1 | NP_001265650.1 | 5, 6 | 681 | |||
NM_001318733.1 | NP_001305662.1 | 5, 6 | 1003 | |||
GenBank mRNA | AJ272344.1 | - | 6, 7 | 866 | ||
BC120874.2 | - | 5, 6 | 995 | 40081041 | ||
BC120875.2 | - | 5, 6 | 996 | 40081042 | ||
BI544995.1 | - | 5, 6 | 669 | |||
Y17108.1 | - | 5, 6 | 1003 |