Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC28A1 SIRNA Assays ›
Gene Symbol:
SLC28A1
Gene Name:
solute carrier family 28 member 1
Gene Aliases:
CNT1, HCNT1
Chromosome Location:
Chr.15: 84884351 - 84975649 on Build GRCh38
UniGene ID:
Hs.459187
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s194991
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC28A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287761.1
NP_001274690.1
5
443
NM_001287762.1
NP_001274691.1
5
443
NM_001321721.1
NP_001308650.1
6
559
NM_001321722.1
NP_001308651.1
6
559
NM_004213.4
NP_004204.3
6
559
NM_201651.2
NP_964014.1
6
559
XM_011522203.2
XP_011520505.1
6
542
XM_011522204.2
XP_011520506.1
7
1062
XM_011522205.2
XP_011520507.1
6
946
XM_011522206.2
XP_011520508.1
5
745
XM_011522207.2
XP_011520509.1
5
387
XM_011522208.2
XP_011520510.1
4
664
XM_011522209.2
XP_011520511.1
6
542
XM_011522210.2
XP_011520512.1
6
542
XM_011522211.2
XP_011520513.1
5
777
XM_011522212.1
XP_011520514.1
6
542
XM_011522214.2
XP_011520516.1
6
542
XM_011522215.2
XP_011520517.1
6
542
XM_011522216.2
XP_011520518.1
4
298
XM_011522217.1
XP_011520519.1
6
542
XM_011522218.2
XP_011520520.1
6
542
XM_017022723.1
XP_016878212.1
6
542
GenBank mRNA
AF309632.1
-
6
538
AK291997.1
-
6
559
AK296074.1
-
6
558
AK296075.1
-
5
443
AK296119.1
-
5
468
AK298369.1
-
6
559
BC029788.1
-
6
519
BC039898.1
-
5
416
BC126204.1
-
5
449
8991920
BC126206.1
-
5
449
8991922
BC143908.1
-
5
448
U62967.1
-
4
351
U62968.1
-
4
351
Back To Top
More Information
Panther Classification:
Molecular Function -
transporter
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
pyrimidine nucleobase transport
nucleoside transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
purine nucleobase transmembrane transport
nucleoside transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0