Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080701.3 | NP_542432.2 | 2 | 728 | ||
GenBank mRNA | AF151107.1 | - | 1 | 184 |