Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPREB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013378.2 | NP_037510.1 | 2 | 282 | ||
GenBank mRNA | AF163825.1 | - | 2 | 244 | ||
AK312098.1 | - | 2 | 237 | |||
AY359000.1 | - | 2 | 282 | |||
BC020666.1 | - | 2 | 307 | 4772032 | ||
BQ899182.1 | - | 2 | 215 | |||
CR456610.1 | - | 2 | 231 |