Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005490.2 | NP_005481.2 | 3 | 468 | ||
XM_005259467.1 | XP_005259524.1 | 3 | 418 | |||
XM_006722602.1 | XP_006722665.1 | 3 | 418 | |||
XM_011527608.2 | XP_011525910.1 | 2 | 835 | |||
XM_011527609.2 | XP_011525911.1 | 2 | 332 | |||
XM_017026123.1 | XP_016881612.1 | 3 | 418 | |||
XM_017026124.1 | XP_016881613.1 | 3 | 418 | |||
GenBank mRNA | AF124249.1 | - | 3 | 425 | ||
AK024368.1 | - | 2 | 1107 | |||
AK225386.1 | - | 3 | 391 | |||
AK290293.1 | - | 3 | 544 | |||
AK292781.1 | - | 3 | 391 | |||
AK310659.1 | - | 3 | 391 | |||
AY358406.1 | - | 3 | 425 | |||
BC006281.2 | - | 3 | 405 | 3958342 |