Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280557.1 | NP_001267486.1 | 3 | 395 | ||
NM_017767.2 | NP_060237.2 | 10 | 2040 | |||
NM_130849.3 | NP_570901.2 | 11 | 1896 | |||
XM_006716599.1 | XP_006716662.1 | 10 | 1722 | |||
GenBank mRNA | AK000489.1 | - | 3 | 390 | ||
AK000695.1 | - | 10 | 1955 | |||
AK025537.1 | - | 11 | 1890 | |||
AK056900.1 | - | 3 | 389 | |||
AK290563.1 | - | 11 | 1844 | |||
BC001688.2 | - | 3 | 367 | |||
BC062625.1 | - | 11 | 1876 | 5203858 | ||
BE798833.1 | - | 2 | 528 | |||
KU178707.1 | - | 11 | 1790 | |||
KU178708.1 | - | 10 | 1508 |