Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019104.2 | NP_061977.1 | 5, 6 | 650 | ||
GenBank mRNA | AF162447.1 | - | 14 | |||
AK293116.1 | - | 5, 6 | 415 | |||
AK301992.1 | - | 5, 6 | 650 | |||
BC009071.1 | - | 5, 6 | 369 | 3872616 | ||
BI767008.1 | - | 5, 6 | 400 |