Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020433.4 | NP_065166.2 | 1 | 995 | ||
NM_175913.3 | NP_787109.2 | 1 | 995 | |||
XM_006723833.3 | XP_006723896.1 | 1 | 987 | |||
GenBank mRNA | AL132999.1 | - | 1 | 510 | ||
BC029957.1 | - | 1 | 995 |