Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012288.2 | NP_001012288.2 | 7 | 980 | ||
NM_022148.3 | NP_071431.2 | 8 | 1083 | |||
NR_110830.1 | 8 | 1262 | ||||
XM_011546181.2 | XP_011544483.1 | 8 | 1118 | |||
GenBank mRNA | AB052639.1 | - | 8 | 994 | ||
AF142570.1 | - | 8 | 1010 | |||
AF338733.1 | - | 8 | 994 | |||
AK026800.1 | - | 8 | 1264 | |||
AK223303.1 | - | 8 | 1110 | |||
AK225583.1 | - | 8 | 1264 | |||
AY775789.1 | - | 7 | 891 |