Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030578.3 | NP_085055.2 | 2 | 280 | ||
XM_011527349.2 | XP_011525651.1 | 2 | 231 | |||
GenBank mRNA | BC004157.2 | - | 2 | 239 | 2819388 | |
BC004444.1 | - | 2 | 239 | 2819388 | ||
BM833644.1 | - | 2 | 253 |