Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136044.1 | NP_001129516.1 | 3 | 1310 | ||
NM_031434.3 | NP_113622.1 | 3 | 1773 | |||
GenBank mRNA | AY037155.1 | - | 3 | 1291 | ||
AY358481.1 | - | 3 | 1269 | |||
AY603380.1 | - | 2 | 1295 | |||
BC000936.1 | - | 3 | 1310 | 3449979 | ||
BC033182.1 | - | 3 | 1448 | 4586932 |