Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032144.2 | NP_115520.2 | 1 | 1216 | ||
GenBank mRNA | AF309646.1 | - | 1 | 809 | ||
AK055504.1 | - | 2 | 603 | |||
AL136727.1 | - | 1 | 1217 | |||
BC120999.1 | - | 1 | 1054 | 40116526 |