Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040160.2 | NP_001035250.1 | 5 | 603 | ||
NM_001040161.2 | NP_001035251.1 | 3 | 380 | |||
NM_001040162.2 | NP_001035252.1 | 4 | 440 | |||
NM_001040165.2 | NP_001035255.1 | 4 | 543 | |||
NM_001288710.1 | NP_001275639.1 | 5 | 611 | |||
NM_032366.4 | NP_115742.3 | 6 | 671 | |||
NR_109976.1 | 5 | 615 | ||||
NR_109977.1 | 5 | 508 | ||||
NR_109978.1 | 6 | 778 | ||||
NR_109979.1 | 4 | 448 | ||||
XM_011522713.2 | XP_011521015.1 | 6 | 870 | |||
XM_011522714.2 | XP_011521016.1 | 5 | 695 | |||
GenBank mRNA | AI146806.1 | - | 4 | 437 | ||
AK057077.1 | - | 3 | 1569 | |||
AK294482.1 | - | 6 | 607 | |||
AK295628.1 | - | 5 | 539 | |||
BC007207.1 | - | 3 | 362 | 4054799 | ||
BI762987.1 | - | 6 | 647 | |||
BM827705.1 | - | 4 | 375 | |||
BP276058.1 | - | 5 | 570 | |||
BQ921757.1 | - | 576 | ||||
BU539308.1 | - | 4 | 396 |