Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005499.2 | NP_005490.1 | 9 | 905 | ||
XM_005258404.3 | XP_005258461.2 | 10 | 1186 | |||
XM_006722962.2 | XP_006723025.1 | 8 | 796 | |||
XM_011526304.2 | XP_011524606.1 | 10 | 1186 | |||
XM_017026134.1 | XP_016881623.1 | 10 | 1186 | |||
GenBank mRNA | AF079566.1 | - | 9 | 925 | ||
AF090384.1 | - | 9 | 871 | |||
AF110957.1 | - | 9 | 835 | |||
AK023120.1 | - | 9 | 906 | |||
AK024114.1 | - | 1 | 80 | |||
AK124730.1 | - | 9 | 1046 | |||
AK299514.1 | - | 6 | 689 | |||
AK315521.1 | - | 9 | 882 | |||
AL136905.1 | - | 9 | 860 | |||
BC003153.1 | - | 9 | 883 | 2905977 | ||
BT009781.1 | - | 9 | 835 | |||
CR456756.1 | - | 9 | 835 | |||
U35832.1 | - | 9 | 860 |