Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011724.1 | NP_001011724.1 | 7 | 1991 | ||
NM_001011725.1 | NP_001011725.1 | 6 | 1850 | |||
GenBank mRNA | AK091082.1 | - | 1 | 1703 | ||
AK094423.1 | - | 6 | 1741 | |||
AK123159.1 | - | 5 | 1530 | |||
AK126454.1 | - | 1 | 2625 | |||
BC082242.1 | - | 6 | 1781 | |||
BC108266.1 | - | 1 | 979 | 4778431 |
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC121133.2 | - | 8, 9 | 962 | 40120894 |