Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152533.2 | NP_689746.1 | 4 | 878 | ||
XM_017005751.1 | XP_016861240.1 | 4 | 848 | |||
XM_017005752.1 | XP_016861241.1 | 4 | 710 | |||
GenBank mRNA | BC032025.1 | - | 4 | 828 | 5165128 |