Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077503.1 | NP_001070971.1 | 4 | 404 | ||
NM_001077504.1 | NP_001070972.1 | 4 | 377 | |||
NM_001077505.1 | NP_001070973.1 | 3 | 344 | |||
NM_001077506.1 | NP_001070974.1 | 5 | 437 | |||
NM_170685.2 | NP_733786.2 | 5 | 455 | |||
GenBank mRNA | AF515828.1 | - | 5 | 455 | ||
AF515829.1 | - | 4 | 404 | |||
AF515830.1 | - | 4 | 377 | |||
AF515831.1 | - | 3 | 344 | |||
AY471574.1 | - | 5 | 437 |