Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147199.3 | NP_671732.3 | 1 | 476 | ||
GenBank mRNA | BC096835.1 | - | 1 | 484 | 30706612 |
Set Membership: |
G-protein coupled receptor |