Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST2H3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005464.2 | NP_001005464.1 | 1 | 388 |
Gene Symbol: HIST2H3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021059.2 | NP_066403.2 | 1 | 388 | ||
GenBank mRNA | BC015544.1 | - | 1 | 388 | ||
BC074969.2 | - | 1 | 380 | 30915509 | ||
BC130635.1 | - | 1 | 382 | 40146666 | ||
BC130637.1 | - | 1 | 382 | 40146668 |