Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039842.2 | NP_001034931.1 | 2 | 614 | ||
NM_001304994.1 | NP_001291923.1 | 2 | 968 | |||
NM_001304995.1 | NP_001291924.1 | 2 | 855 | |||
NM_001304999.1 | NP_001291928.1 | 1 | 1547 | |||
NR_130936.1 | 2 | 977 | ||||
NR_130937.1 | 2 | 982 | ||||
NR_130938.1 | 2 | 986 | ||||
NR_130939.1 | 2 | 643 | ||||
GenBank mRNA | AY007126.1 | - | 1 | 1545 | ||
BC090923.1 | - | 2 | 565 | 30335041 | ||
CB048026.1 | - | 2 | 651 |