Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267868.1 | NP_001254797.1 | 9 | 2068 | ||
NM_001267869.1 | NP_001254798.1 | 6 | 1755 | |||
NM_001267870.1 | NP_001254799.1 | 5 | 1595 | |||
NM_001267871.1 | NP_001254800.1 | 6 | 1524 | |||
NM_198533.2 | NP_940935.1 | 8 | 1767 | |||
NM_198704.2 | NP_941993.1 | 6 | 1592 | |||
NM_198705.2 | NP_941994.1 | 6 | 1687 | |||
NM_198706.2 | NP_941995.1 | 8 | 1930 | |||
NM_198707.2 | NP_941996.1 | 7 | 1867 | |||
NM_198708.2 | NP_941997.1 | 7 | 1818 | |||
GenBank mRNA | AY268351.1 | - | 6 | 1428 | ||
AY268352.1 | - | 8 | 1631 | |||
AY268353.1 | - | 7 | 1591 | |||
AY268354.1 | - | 7 | 1519 | |||
AY313894.1 | - | 8 | 1496 | |||
AY313895.1 | - | 6 | 1333 | |||
AY313896.1 | - | 6 | 1265 | |||
BC053546.1 | - | 7 | 1448 | |||
BC072434.1 | - | 6 | 1504 | |||
BC094798.1 | - | 6 | 1511 |