Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163724.2 | NP_001157196.1 | 4 | 458 | ||
NM_001288583.1 | NP_001275512.1 | 4 | 431 | |||
GenBank mRNA | BC035379.1 | - | 3 | 406 | ||
BC042129.1 | - | 4 | 457 | |||
BC146945.1 | - | 2 | 263 | |||
BC146957.1 | - | 2 | 263 | |||
HY003308.1 | - | 4 | 458 | |||
HY032469.1 | - | 4 | 431 | |||
KC152644.1 | - | 4 | 422 |