Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4N2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004723.2 | NP_001004723.1 | 2 | 347 | ||
GenBank mRNA | BM696516.1 | - | 2 | 347 |