Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168412.1 | NP_001161884.1 | 3 | 461 | ||
NM_005724.5 | NP_005715.1 | 4 | 653 | |||
NM_198902.2 | NP_944492.1 | 3 | 578 | |||
XM_017021857.1 | XP_016877346.1 | 4 | 262 | |||
GenBank mRNA | AF054840.1 | - | 4 | 379 | ||
AF133423.1 | - | 4 | 453 | |||
AK001305.1 | - | 4 | 499 | |||
AK001310.1 | - | 4 | 485 | |||
AK001326.1 | - | 4 | 596 | |||
AK027793.1 | - | 1 | 1704 | |||
AK075336.1 | - | 4 | 534 | |||
AK075482.1 | - | 3 | 459 | |||
AK293677.1 | - | 5 | 442 | |||
AK298157.1 | - | 3 | 342 | |||
BC000704.2 | - | 4 | 554 | 3349181 | ||
BC004280.1 | - | 4 | 515 | 3616380 | ||
BC009248.2 | - | 4 | 518 | 2964568 | ||
BC011206.1 | - | 4 | 547 | 3457158 |