Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005731.3 | NP_005722.1 | 3 | 220 | ||
NM_152862.2 | NP_690601.1 | 4 | 315 | |||
XM_017003113.1 | XP_016858602.1 | 3 | 189 | |||
XM_017003114.1 | XP_016858603.1 | 4 | 262 | |||
XM_017003115.1 | XP_016858604.1 | 4 | 225 | |||
GenBank mRNA | AF006085.1 | - | 3 | 230 | ||
AF279893.1 | - | 2 | 72 | |||
AK290268.1 | - | 4 | 214 | |||
BC000590.2 | - | 4 | 178 | 3346685 | ||
BG282181.1 | - | 4 | 314 | |||
BG765898.1 | - | 4 | 258 | |||
BT006898.1 | - | 3 | 146 | |||
U50523.1 | - | 4 | 230 |