Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 8 | 1814 | |||
GenBank mRNA | BC045636.1 | - | 8 | 1817 | ||
BC068610.1 | - | 8 | 1817 | |||
BC110795.1 | - | 4 | 1081 |
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 11 | 2279 | ||
GenBank mRNA | BC056606.1 | - | 11 | 2280 | 4838469 |