Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080524.1 | NP_001073993.1 | 3 | 510 | ||
XM_005255502.3 | XP_005255559.1 | 3 | 576 |