Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400493.1 | - | LIT2215 IC-SNURF-SNRPN 3 prime noncoding transcripts |
2, 3 | 169 | |
AY362864.1 | - | 5, 6 | 454 |