Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AF400493.1 | - | LIT2215 IC-SNURF-SNRPN 3 prime noncoding transcripts |
3 | 192 | |
AF400494.1 | - | LIT2208 IC-SNURF-SNRPN 3 prime noncoding transcripts |
1 | 13 | ||
AY362864.1 | - | 6 | 477 |