Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 10 | 980 | ||
NM_001291398.1 | NP_001278327.1 | 10 | 980 | |||
XM_005269806.3 | XP_005269863.2 | 10 | 928 | |||
XM_005269807.3 | XP_005269864.2 | 10 | 928 | |||
XM_005269808.3 | XP_005269865.2 | 10 | 928 | |||
XM_006717831.3 | XP_006717894.2 | 10 | 928 | |||
XM_006717832.3 | XP_006717895.2 | 10 | 928 | |||
XM_011539786.2 | XP_011538088.1 | 10 | 928 | |||
XM_017016227.1 | XP_016871716.1 | 10 | 928 | |||
XM_017016228.1 | XP_016871717.1 | 10 | 928 | |||
XM_017016229.1 | XP_016871718.1 | 10 | 928 | |||
XM_017016230.1 | XP_016871719.1 | 9 | 793 | |||
XM_017016231.1 | XP_016871720.1 | 10 | 928 | |||
XM_017016232.1 | XP_016871721.1 | 10 | 928 | |||
XM_017016233.1 | XP_016871722.1 | 7 | 503 | |||
XM_017016234.1 | XP_016871723.1 | 11 | 1022 | |||
XM_017016235.1 | XP_016871724.1 | 2 | 49 | |||
GenBank mRNA | AK001686.1 | - | 7 | 631 | ||
AK295129.1 | - | 10 | 932 | |||
AK304116.1 | - | 10 | 916 | |||
AK311628.1 | - | 7 | 534 | |||
BC075815.1 | - | 7 | 545 | |||
BC082258.1 | - | 10 | 907 | 6278433 | ||
BC144479.1 | - | 10 | 1009 | 9053006 | ||
DA402580.1 | - | 2 | 138 | |||
DC403380.1 | - | 7 | 534 |