Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010110.1 | XP_016865599.1 | 5 | 534 | ||
XM_017010111.1 | XP_016865600.1 | 4 | 361 | |||
XM_017010112.1 | XP_016865601.1 | 4 | 938 | |||
XM_017010113.1 | XP_016865602.1 | 5 | 1060 | |||
XM_017010114.1 | XP_016865603.1 | 5 | 3821 | |||
XM_017010115.1 | XP_016865604.1 | 4 | 886 | |||
XM_017010116.1 | XP_016865605.1 | 3 | 1286 | |||
XM_017010117.1 | XP_016865606.1 | 3 | 1286 | |||
XM_017010118.1 | XP_016865607.1 | 3 | 1286 | |||
XM_017010119.1 | XP_016865608.1 | 3 | 1286 | |||
XM_017010120.1 | XP_016865609.1 | 3 | 1286 | |||
XM_017010121.1 | XP_016865610.1 | 3 | 1286 | |||
XM_017010122.1 | XP_016865611.1 | 3 | 1286 | |||
XM_017010123.1 | XP_016865612.1 | 3 | 1286 | |||
XM_017010124.1 | XP_016865613.1 | 5 | 534 |