Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 11 | 1304 | ||
XM_005256769.1 | XP_005256826.1 | 11 | 1301 | |||
XM_005256770.1 | XP_005256827.1 | 10 | 1205 | |||
XM_011523993.1 | XP_011522295.1 | 10 | 1208 | |||
XM_011523994.1 | XP_011522296.1 | 9 | 1172 | |||
XM_011523995.1 | XP_011522297.1 | 11 | 1216 | |||
XM_011523996.1 | XP_011522298.1 | 10 | 1208 | |||
XM_011523997.1 | XP_011522299.1 | 9 | 942 | |||
GenBank mRNA | AK315818.1 | - | 11 | 1291 | ||
BC073889.1 | - | 11 | 1273 | 5222492 |