Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042705.2 | NP_001036170.1 | 3 | 319 | ||
NM_001042706.2 | NP_001036171.1 | 3 | 319 | |||
GenBank mRNA | BC137464.1 | - | 3 | 199 | 9021464 | |
BC137469.1 | - | 3 | 199 | 9021469 | ||
DQ309553.1 | - | 3 | 243 | |||
DQ309554.1 | - | 3 | 243 |
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 3 | 319 | ||
GenBank mRNA | AK096479.1 | - | 3 | 225 | ||
DQ157848.1 | - | 2 | 138 |