Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278495.1 | NP_001265424.1 | 7 | 2811 |
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099338.1 | NP_001092808.1 | 7 | 2811 | ||
XM_011540122.2 | XP_011538424.1 | 9 | 5654 | |||
GenBank mRNA | BC092519.1 | - | 7 | 2807 |