Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP19-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099219.1 | NP_001092689.1 | 1 | 227 | ||
GenBank mRNA | AB096964.1 | - | 1 | 227 |