Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM90A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319982.1 | NP_001306911.1 | 3, 4 | 529 | ||
NM_018088.3 | NP_060558.3 | 4, 5 | 686 | |||
XM_011520719.1 | XP_011519021.1 | 4, 5 | 677 | |||
XM_017019547.1 | XP_016875036.1 | 4, 5 | 674 | |||
GenBank mRNA | AK001270.1 | - | 3, 4 | 512 | ||
BC042608.1 | - | 4, 5 | 665 | 5167203 |
Gene Symbol: FAM90A25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832996.1 | - | 1, 2 | 69 |
Gene Symbol: FAM90A2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046354.1 | 2, 3 | 215 | |||
GenBank mRNA | CX762572.1 | - | 2, 3 | 251 |