Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126340.2 | NP_001119812.1 | 4 | 1447 | ||
NM_001271818.1 | NP_001258747.1 | 4 | 1470 | |||
NM_001271819.1 | NP_001258748.1 | 3 | 1209 | |||
NM_032831.3 | NP_116220.1 | 3 | 1338 | |||
GenBank mRNA | AK022536.1 | - | 1 | 279 | ||
AK027639.1 | - | 3 | 1292 | |||
AK056497.1 | - | 4 | 1402 | |||
BC069270.1 | - | 3 | 1241 | 5477442 | ||
CR457287.1 | - | 1 | 254 | |||
Y10777.1 | - | 1 | 408 |