Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261390.1 | NP_001248319.1 | 4 | 393 | ||
NM_001261391.1 | NP_001248320.1 | 4 | 384 | |||
NM_001261393.1 | NP_001248322.1 | 3 | 321 | |||
NM_001261395.1 | NP_001248324.1 | 2 | 131 | |||
NM_005831.4 | NP_005822.1 | 3 | 321 | |||
GenBank mRNA | AK222666.1 | - | 3 | 271 | ||
AK223227.1 | - | 3 | 264 | |||
AK293137.1 | - | 2 | 84 | |||
AK293329.1 | - | 4 | 346 | |||
AK298177.1 | - | 4 | 336 | |||
AK303313.1 | - | 3 | 274 | |||
AK304024.1 | - | 3 | 274 | |||
AK314796.1 | - | 3 | 274 | |||
AL042638.1 | - | 3 | 319 | |||
AL046722.1 | - | 3 | 321 | |||
BC004130.2 | - | 3 | 260 | 3505988 | ||
BC015893.2 | - | 2 | 230 | 3921397 | ||
BG773301.1 | - | 3 | 303 | |||
CR456763.1 | - | 2 | 220 | |||
U22897.1 | - | 3 | 274 |