Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000057.3 | NP_000048.1 | 22 | 4224 | ||
NM_001287246.1 | NP_001274175.1 | 23 | 4334 | |||
NM_001287247.1 | NP_001274176.1 | 20 | 3831 | |||
NM_001287248.1 | NP_001274177.1 | 22 | 4390 | |||
XM_006720632.2 | XP_006720695.1 | 16 | 2283 | |||
XM_011521881.2 | XP_011520183.1 | 19 | 3259 | |||
GenBank mRNA | AK295194.1 | - | 21 | 3290 | ||
AK314262.1 | - | 22 | 4153 | |||
BC093622.1 | - | 22 | 4147 | 7939467 | ||
BC101567.1 | - | 22 | 4147 | 8069073 | ||
BC115030.1 | - | 22 | 4172 | 7961413 | ||
BC115032.1 | - | 22 | 4172 | 7961425 | ||
BC143280.1 | - | 22 | 4312 | 9051783 | ||
DB520927.1 | - | 2 | 132 | |||
U39817.1 | - | 22 | 4180 |