Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304990.1 | NP_001291919.1 | 3 | 592 | ||
NM_005840.2 | NP_005831.1 | 2 | 736 | |||
GenBank mRNA | AK125483.1 | - | 2 | 661 | ||
AK289543.1 | - | 2 | 729 | |||
BC069609.1 | - | 1 | 298 | 7262284 | ||
BC074972.2 | - | 1 | 274 | 30915293 | ||
BC074973.2 | - | 1 | 274 | 30915511 |