Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297647.1 | NP_001284576.1 | 3 | 336 | ||
NM_001297648.1 | NP_001284577.1 | 3 | 336 | |||
NM_006314.2 | NP_006305.2 | 3 | 336 | |||
XM_011540484.2 | XP_011538786.1 | 3 | 316 | |||
GenBank mRNA | AB209335.1 | - | 3 | 331 | ||
AF100153.1 | - | 3 | 314 | |||
AK001900.1 | - | 3 | 315 | |||
AK222904.1 | - | 3 | 319 | |||
AK292646.1 | - | 3 | 318 | |||
AK296814.1 | - | 3 | 336 | |||
AK302056.1 | - | 3 | 317 | |||
BC011604.2 | - | 3 | 317 | 3139227 | ||
BC012797.2 | - | 3 | 317 | 3505293 | ||
BT006900.1 | - | 3 | 278 | |||
DA445069.1 | - | 3 | 336 | |||
DA868484.1 | - | 3 | 317 | |||
DR159101.1 | - | 3 | 279 |